Learn More
MSC GOJO Healthcare Surface Disinfectant, 1 Gallon, Fragrance Free, Ready-to-Use
Catalog No. 502837287
Encompass
This disinfectant is a 1-gallon, fragrance-free, ethyl alcohol-based solution designed for healthcare surface disinfection. It is effective against a wide range of pathogens on hard and soft surfaces and is EPA Design for the Environment certified.
  • Kills human coronavirus in 10 seconds.
  • Achieves 30-second disinfection for MRSA, VRE, and Norovirus.
  • Fragrance-free formula contains no bleach or harsh fumes.
  • Requires no rinsing on food contact surfaces.
  • Proven effective across most hard and soft surfaces.
  • Formulated with ethyl alcohol as the active ingredient.
  • Awarded the U.S. EPA Design for the Environment certification (DfE).
  • Achieved the EPA's lowest toxicity rating.
  • Safe to use around children and pregnant women.
Catalog No. 50-283-7287 Supplier MSC Supplier No. 37643541
This disinfectant is a 1-gallon, fragrance-free, ethyl alcohol-based solution designed for healthcare surface disinfection. It is effective against a wide range of pathogens on hard and soft surfaces and is EPA Design for the Environment certified.
  • Kills human coronavirus in 10 seconds.
  • Achieves 30-second disinfection for MRSA, VRE, and Norovirus.
  • Fragrance-free formula contains no bleach or harsh fumes.
  • Requires no rinsing on food contact surfaces.
  • Proven effective across most hard and soft surfaces.
  • Formulated with ethyl alcohol as the active ingredient.
  • Awarded the U.S. EPA Design for the Environment certification (DfE).
  • Achieved the EPA's lowest toxicity rating.
  • Safe to use around children and pregnant women.