Learn More
Description
- Refill for PURELL™ CS8 Touch-Free Waterless Surgical Scrub Dispenser
- Contains five skin-conditioning agents that leave hands feeling soft and moisturized without the feeling of stickiness
- Quick and easy-requires no water or scrub brush
- Compatible with latex, nitrile, neoprene and polyisoprene gloves, dries quickly for easy gloving
- Chlorhexidine Gluconate (CHG) compatible formulation
- Creates a pH-balanced environment that helps inhibit germ recovery and regrowth
- Ideal for high acuity areas
- Hypoallergenic, dermatologist tested, dye free, fragrance free
- Energy-on-the-refill technology - every refill has a built-in energy source, designed to last as long as the refill
- SANITARY SEALED™ Collapsible PET refill is easily recycled
Specifications
Specifications
| Product Type | Hand Sterilizer/Disinfectant |
| Color | Clear |
| Form | Gel |
| Size | 1200 mL |
| Volume (Metric) | 1200 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
