Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Healthy Plants
SCI_ED

Catalog No. S02070 Shop All Aldon Products
Change view
Click to view available options
Grade:
K to 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02070 K to 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING CHOKING HAZARD Small parts. Not for children under 3 yrs. // WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, which are known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.
Catalog No. S02070 Supplier Innovating Science™ Supplier No. IS1005
Add to Cart
Add to Cart
WARNING CHOKING HAZARD Small parts. Not for children under 3 yrs. // WARNING: This product can expose you to chemicals including Strong inorganic acid mists containing sulfuric acid, which are known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.

Students will examine and learn the needs of plants for growth and survival by growing bean seeds in a variety of conditions.

Designed for older elementary students to help them examine and understand the needs of plants for growth and survival. For the sake of time, the class can be split in thirds, with some students conducting a light/dark experiment, some studying plant nutritional needs, and some studying effects of acid rain. Results will be shared at the end. This kit contains materials for 15 groups of two.

Specifications

Grade K to 5
Class Size 15 Lab Groups
Product Type Botany Classroom Kits
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.