Learn More
3B Scientific™ Heart & Breath Sounds Simulator
SDPSCI_ED
Catalog No. S00271 Shop All 3B Scientific Products
Change view
Click to view available options
Includes:
Two External Sounders
Catalog No. Includes
S00271 Two External Sounders
1 options
This item is not returnable. View return policy
Catalog No. S00271 Supplier 3B Scientific™ Supplier No. 1005884
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Heart and Breath Sounds Simulator unit includes two external sounders with which students can auscultate the sounds with a stethoscope.
  • This hand-held device simulates both heart and breath sounds and allows the determination of these by plugging in one of 7 different menus, each containing 16 different sounds.
  • The Heart and Breath Sounds Simulator unit includes two external sounders with which students can auscultate the sounds with a stethoscope.
  • Linking additional stethoscope sounders (see W49430), up to 15 students can auscultate at the same time. A Loud Speaker may also be connected.
  • Additionally, the unit has a volume control and two LED's that illuminate during systole (heart menu) and expiration (breath menu).
  • Virtually maintenance-free, the unit is powered by a 9 volt battery.
  • All data used in the menus are actual patient sounds. Sounds of the same name are from different patients in different menus to show appearance variations.
  • Cartridges with sounds required as well and not included in this item.
Specifications
Product Type Sound Simulator
Dimensions (L x W x H) 3 x 2.4 x 2.2 in.
Includes Two External Sounders
Weight (English) 1.24 lb.