Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Heart Infusion Broth, 500 Grams
SDP

Catalog No. 501368292
Encompass_Preferred

Recommended for the cultivations of rare microorganisms. Protect from moisture and light.

Brain Heart Infusion from solids: 2.0g
Casein Digest Peptone: 10.0g
Pepetic Digest of animal tissue: 5.0g
Yeast Extract: 3.0g
Sodium Chloride: 5.0g

Catalog No. 50-136-8292 Supplier Research Products International Corp Supplier No. H11000500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recommended for the cultivations of rare microorganisms. Protect from moisture and light.

Brain Heart Infusion from solids: 2.0g
Casein Digest Peptone: 10.0g
Pepetic Digest of animal tissue: 5.0g
Yeast Extract: 3.0g
Sodium Chloride: 5.0g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.