Learn More
MOLTOX Molecular Toxicology Inc Heart Infusion Agar w/ Congo Red & D-galactose, 20 mL/plate, 10 plates/sleeve, Store in Refrigerator
SDP
Catalog No. 50948634
Encompass_Preferred
Heart Infusion agar is a general purpose medium used for the cultivation of a wide variety of fastidious organisms and specifically for the isolation of Vibrio species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-634 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 2140S142
Heart Infusion agar is a general purpose medium used for the cultivation of a wide variety of fastidious organisms and specifically for the isolation of Vibrio species. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.