Learn More
ProSci Heartland Virus Glycoprotein 2 Peptide

Description
Specifications
Specifications
| Species | Heartland Virus |
| Accession Number | AFP33393 |
| Common Name | Heartland Virus Glycoprotein 2 |
| Concentration | 200 μg/mL |
| Content And Storage | Store Heartland Virus Glycoprotein 2 peptide (at -20°C, stable for one year.) |
| Format | Liquid |
| Formulation | PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide |
| For Use With (Application) | Blocking Assay |
| Quantity | 0.05 mg |
| Sequence | 19 amino acids near the center of Heartland Virus Glycoprotein 2. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.