Learn More
Description
This peptide corresponds to 19 amino acids near the center of Heartland Virus Glycoprotein 2. For use as a blocking peptide in Western blot, incubate the peptide with equal volume of antibody for 30 min at 37°C.
Specifications
Specifications
| Species | Virus |
| Purification Method | purified |
| Common Name | Heartland Virus Glycoprotein 2 |
| Concentration | 200 μg/mL |
| Content And Storage | -20°C |
| Description | Synthetic Peptide |
| Format | Liquid |
| Formulation | PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 |
| For Use With (Application) | Blocking Assay, Control |
| Lyophilized | No |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.