Learn More
World Precision Instrument Heat Controller/Humidity Monitor -AIRTHERM-SMT-1W
SDP
Supplier:  World Precision Instrument AIRTHERMSMT1W
Encompass
The AirTherm™ SMT is a new generation of heat control system from WPI designed to be used in Live Cell Imaging applications with microscopes fitted with a full microscope environmental chamber enclosure. The standard AirTherm™ SMT controls temperature and, as an option, monitors humidity.
To learn more about our warranty options, click here.
Catalog No. 50-253-8932
Only null left
Add to Cart
Request a Quote