Learn More
World Precision Instrument Heat Controller/Humidity Monitor -AIRTHERM-SMT-2W
SDP
Catalog No. 502538931
Encompass
The AirTherm™ SMT is a new generation of heat control system from WPI designed to be used in Live Cell Imaging applications with microscopes fitted with a full microscope environmental chamber enclosure. The standard AirTherm™ SMT controls temperature and, as an option, monitors humidity.
To learn more about our warranty options, click here.
Catalog No. 50-253-8931 Supplier World Precision Instrument Supplier No. AIRTHERMSMT2W
Add to Cart
Add to Cart
Request a Quote
The AirTherm™ SMT is a new generation of heat control system from WPI designed to be used in Live Cell Imaging applications with microscopes fitted with a full microscope environmental chamber enclosure. The standard AirTherm™ SMT controls temperature and, as an option, monitors humidity.
To learn more about our warranty options, click here.