Learn More
Bellco Glass, Inc. Heat/Cool Coil,15L
SDP
Catalog No. 501944800
Encompass_Preferred
Heating and cooling coils are used for cell culture temperature manipulation within the bioreactor vessel. Must be hooked up to a water circulating mechanism for heating or cooling. Fits through ports on a stainless steel head plate. MADE IN THE USA. 316l electropolished. FOR CELL CULTURE TEMPERATURE MANIPULATION. OD X OAL: 3 1/4 X 16 1/2". #TURNS: 5 1/2.
Catalog No. 50-194-4800 Supplier Bellco Glass, Inc. Supplier No. 196450115
Heating and cooling coils are used for cell culture temperature manipulation within the bioreactor vessel. Must be hooked up to a water circulating mechanism for heating or cooling. Fits through ports on a stainless steel head plate. MADE IN THE USA. 316l electropolished. FOR CELL CULTURE TEMPERATURE MANIPULATION. OD X OAL: 3 1/4 X 16 1/2". #TURNS: 5 1/2.