Learn More
Grainger Heat Detector Coverage 1600 Sq. Ft.
Catalog No. 502765741
Encompass_Preferred
The Grainger heat detectors are designed to activate an alarm upon detecting an abnormally high temperature. They use a fixed temperature or rate-of-rise detection method to identify fires. These devices are UL listed and FM approved, providing reliable fire detection. The heat detectors come in multiple styles including ceiling-mount, wall-mount and duct-mount options. Key features include LED operation indicators, low-profile designs and wide fields of detection. Overall, these heat detectors offer technologically advanced, compliant fire detection critical for protecting facilities.
Catalog No. 50-276-5741 Supplier Grainger Supplier No. 16X275
The Grainger heat detectors are designed to activate an alarm upon detecting an abnormally high temperature. They use a fixed temperature or rate-of-rise detection method to identify fires. These devices are UL listed and FM approved, providing reliable fire detection. The heat detectors come in multiple styles including ceiling-mount, wall-mount and duct-mount options. Key features include LED operation indicators, low-profile designs and wide fields of detection. Overall, these heat detectors offer technologically advanced, compliant fire detection critical for protecting facilities.