Learn More
Grainger Heat Detector Coverage 2500 Sq. Ft.
Catalog No. 502766960
Encompass_Preferred
The Edwards Signaling Heat Detector is a rate-of-rise heat detector that activates its alarm when the temperature rises at a rate exceeding 15°F per minute. It has a fixed temperature heat sensor that activates at 135°F. The heat detector has a low profile design with a white finish and operates on 24VAC/VDC power. It is UL Listed and meets NFPA 72 standards. The heat detector provides open area protection with a coverage range up to 2,500 sq ft.
Catalog No. 50-276-6960 Supplier Grainger Supplier No. 16X273
The Edwards Signaling Heat Detector is a rate-of-rise heat detector that activates its alarm when the temperature rises at a rate exceeding 15°F per minute. It has a fixed temperature heat sensor that activates at 135°F. The heat detector has a low profile design with a white finish and operates on 24VAC/VDC power. It is UL Listed and meets NFPA 72 standards. The heat detector provides open area protection with a coverage range up to 2,500 sq ft.