Learn More
Grainger Heat Detector Silver/White
Catalog No. 502767175
Encompass_Preferred
The Edwards Signaling Heat Detector Rate Compensation is a fixed temperature heat detector designed for open area protection. It has a 135°F/57°C fixed temperature rating and rate compensation that adjusts sensitivity based on rate of temperature rise for reliable fire detection. The detector provides a 360° field of detection coverage. It has a low profile design with a white finish and operates on 24V systems. The detector meets UL 521 and CSFM standards. It can provide open area protection in commercial, industrial, and institutional buildings.
Catalog No. 50-276-7175 Supplier Grainger Supplier No. 16X271
The Edwards Signaling Heat Detector Rate Compensation is a fixed temperature heat detector designed for open area protection. It has a 135°F/57°C fixed temperature rating and rate compensation that adjusts sensitivity based on rate of temperature rise for reliable fire detection. The detector provides a 360° field of detection coverage. It has a low profile design with a white finish and operates on 24V systems. The detector meets UL 521 and CSFM standards. It can provide open area protection in commercial, industrial, and institutional buildings.