Learn More
Grainger Heat Detector Silver/White
Catalog No. 502766127
Encompass_Preferred
The Edwards Signaling Heat Detector Rate Compensation is a fixed temperature heat detector designed for open area protection. It has a 135°F/57°C fixed temperature rating and rate compensation feature that adjusts sensitivity based on rate of temperature rise for reliable fire detection. This UL listed detector provides a coverage area up to 2,500 sq ft per detector. It has a low profile design with insect resistant cover and operates on 24V systems. The heat detector is made of durable ABS plastic in white finish and includes mounting bracket for easy installation.
Catalog No. 50-276-6127 Supplier Grainger Supplier No. 16X274
The Edwards Signaling Heat Detector Rate Compensation is a fixed temperature heat detector designed for open area protection. It has a 135°F/57°C fixed temperature rating and rate compensation feature that adjusts sensitivity based on rate of temperature rise for reliable fire detection. This UL listed detector provides a coverage area up to 2,500 sq ft per detector. It has a low profile design with insect resistant cover and operates on 24V systems. The heat detector is made of durable ABS plastic in white finish and includes mounting bracket for easy installation.