Learn More
Chemglass Life Sciences Heat Exchanger Loop for 7 L Bioreactors - Chemglass CLS-1383-12
SDP
Catalog No. 502733312
Encompass
Loop-style heat exchanger designed for 7 L bioreactors to provide precise temperature control by circulating a heat-transfer fluid through a stainless steel loop. It installs into headplate ports for straightforward replacement or upgrade of jacketed bioreactor systems.
  • Loop-style exchanger for temperature control of bioreactor vessels.
  • Fits two M10 headplate ports for straightforward installation.
  • 316L stainless steel components with electropolished finish for corrosion resistance.
  • Suitable for 5-15 L reactor sizes, optimized for 7 L vessels.
  • Compatible with common bioprocessing heat-exchange fluids.
Catalog No. 50-273-3312 Supplier Chemglass Life Sciences Supplier No. CLS138312
Loop-style heat exchanger designed for 7 L bioreactors to provide precise temperature control by circulating a heat-transfer fluid through a stainless steel loop. It installs into headplate ports for straightforward replacement or upgrade of jacketed bioreactor systems.
  • Loop-style exchanger for temperature control of bioreactor vessels.
  • Fits two M10 headplate ports for straightforward installation.
  • 316L stainless steel components with electropolished finish for corrosion resistance.
  • Suitable for 5-15 L reactor sizes, optimized for 7 L vessels.
  • Compatible with common bioprocessing heat-exchange fluids.