Learn More
MSC Master Appliance Heat Gun, 650 F, 3.8 CFM, 1 ea
Catalog No. NC1653817
Encompass
This Master Appliance heat gun operates at 650 °F with an airflow of 3.8 CFM, providing precise heating for various applications.
  • Adjustable temperature settings
  • Includes bench stand, heat shrink, and solder preform
  • Heating element is replaceable
  • Corded power type with a 6 ft cord
  • Suitable for applications such as removing decals, curing epoxies, melting solder, shrinking tubing, applying adhesive, and drying components
Catalog No. NC1653817 Supplier MSC Supplier No. 89420236
This Master Appliance heat gun operates at 650 °F with an airflow of 3.8 CFM, providing precise heating for various applications.
  • Adjustable temperature settings
  • Includes bench stand, heat shrink, and solder preform
  • Heating element is replaceable
  • Corded power type with a 6 ft cord
  • Suitable for applications such as removing decals, curing epoxies, melting solder, shrinking tubing, applying adhesive, and drying components