Learn More
Chemglass Life Sciences Chemglass Heat Gun, Heavy-Duty, Multiple Use CG-9001-D-01 - 204°C, 120V, 600W
SDP
Catalog No. NC3314586
Encompass_Preferred
Heavy-duty heat gun designed for laboratory and industrial use, delivering a fixed 204°C output and 600W of power for bending plastics, curing adhesives, shrinking tubing, and stripping paint. Sturdy die-cast aluminum housing, ergonomic soft-grip handle, and adjustable stand improve durability and user comfort.
  • Fixed 204°C operating temperature for consistent heating.
  • 600W power with 120V, 60 Hz electrical rating.
  • 27 CFM air volume at 3,400 FPM for efficient work.
  • Die-cast aluminum housing with reinforced adjustable stand.
  • Ergonomic soft-grip handle for reduced fatigue.
  • 6 ft grounded cord for safe operation.
Catalog No. NC3314586 Supplier Chemglass Life Sciences Supplier No. CG9001D01
Heavy-duty heat gun designed for laboratory and industrial use, delivering a fixed 204°C output and 600W of power for bending plastics, curing adhesives, shrinking tubing, and stripping paint. Sturdy die-cast aluminum housing, ergonomic soft-grip handle, and adjustable stand improve durability and user comfort.
  • Fixed 204°C operating temperature for consistent heating.
  • 600W power with 120V, 60 Hz electrical rating.
  • 27 CFM air volume at 3,400 FPM for efficient work.
  • Die-cast aluminum housing with reinforced adjustable stand.
  • Ergonomic soft-grip handle for reduced fatigue.
  • 6 ft grounded cord for safe operation.