Learn More
Chemglass Life Sciences Heat Gun, Heavy-Duty, Multiple Use, Variable Temperature, 38 - 649°C Temp Range, 120V, 60Hz, 14.5A, 1740W
SDP
Catalog No. NC1772530
Encompass_Preferred
Professional, heavy-duty, multiple use variable temperature heat gun has a sturdy die cast housing and adjustable non-slip stand. The lockable variable temperature control allows the user to dial-in any temperature from 38-649¿ C and lock it in. Used to activate adhesives, defrost frozen coils, heat sensitive material, melt solder, soften plastics and more. High 27 CFM at 3,400 FPM, gets tough jobs done faster. New quick-change plug-in heating element with even airflow element for maximum heating efficiency.
Catalog No. NC1772530 Supplier Chemglass Life Sciences Supplier No. CG9003D01
Professional, heavy-duty, multiple use variable temperature heat gun has a sturdy die cast housing and adjustable non-slip stand. The lockable variable temperature control allows the user to dial-in any temperature from 38-649¿ C and lock it in. Used to activate adhesives, defrost frozen coils, heat sensitive material, melt solder, soften plastics and more. High 27 CFM at 3,400 FPM, gets tough jobs done faster. New quick-change plug-in heating element with even airflow element for maximum heating efficiency.