Learn More
Chemglass Life Sciences Heat Gun Holder
SDP
Catalog No. 501531851
Encompass_Preferred
Dispensette S bottletop dispensers are suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents. Extremely easy filling and dispensing. Simple priming by optimized flow channels. Rapid and safe volume adjustment with internal scalloped track.
Catalog No. 50-153-1851 Supplier Chemglass Life Sciences Supplier No. CG9001H10
Dispensette S bottletop dispensers are suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents. Extremely easy filling and dispensing. Simple priming by optimized flow channels. Rapid and safe volume adjustment with internal scalloped track.