Learn More
Caron Products And Services Humidified Incubator, 10 cu. ft., Temp & RH
Catalog No. NC2583594
Encompass_Preferred
10 ft3 (283 L) Humidified Incubator offers temperature (10C above ambient to 70C) and humidity control (ambient to 98% RH). Ideal for a wide range of applications. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.
Catalog No. NC2583594 Supplier Caron Products And Services Supplier No. 7003101
Add to Cart
Add to Cart
Request a Quote
10 ft3 (283 L) Humidified Incubator offers temperature (10C above ambient to 70C) and humidity control (ambient to 98% RH). Ideal for a wide range of applications. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 115V, 60 Hz.