Learn More
ATCC Heat-inactivated SARS-CoV-2 strain 2019-nCoV/USA-WA1/2020 (ATCC VR-1986HK™)
Catalog No. 502384541
Encompass
Heat-inactivated SARS-CoV-2 strain 2019-nCoV/USA-WA1/2020 (ATCC VR-1986HK™) is a whole-genome sequenced preparation inactivated by heating to 65°C for 30 minutes. Originally isolated from a confirmed human case in Washington state, it is intended for use in molecular assays or as a process control (eg, viral RNA extraction through qPCR amplification process), and it can also be used to make a dilution series to estimate level of detection. Each vial contains ~0.25 mL of clarified cell lysate and supernatant from infected Vero E6 cells.
Catalog No. 50-238-4541 Supplier ATCC Supplier No. VR1986HK
Heat-inactivated SARS-CoV-2 strain 2019-nCoV/USA-WA1/2020 (ATCC VR-1986HK™) is a whole-genome sequenced preparation inactivated by heating to 65°C for 30 minutes. Originally isolated from a confirmed human case in Washington state, it is intended for use in molecular assays or as a process control (eg, viral RNA extraction through qPCR amplification process), and it can also be used to make a dilution series to estimate level of detection. Each vial contains ~0.25 mL of clarified cell lysate and supernatant from infected Vero E6 cells.