Learn More
ATCC Heat inactivated; SARS-CoV-2; variant B.1.1.7
Supplier:  ATCC VR3326HK
Encompass
VR-3326HK is a preparation of SARS-CoV-2 strain USA/CA_CDC_5574/2020 (Alpha variant) that has been inactivated by heating. The strain was isolated in December 2020 from a nasopharyngeal swab from a human in San Diego, California, and was deposited by the Centers for Disease Control and Prevention. This heat-inactivated preparation is whole-genome sequenced and can be used as a process control in the devleopment of SARS-CoV-2 molecular diagnostic tools and can also be used to make a dilution series to estimate the level of detection. Due to high demand, this product is limited to 20 per organization per 60 days.
Catalog No. 50-238-2473