Learn More
V&P Scientific Inc HEAT TRANSFER PLATE, 96 Well PCR Plates, Sculpted
Catalog No. NC9882359
Encompass
HEAT TRANSFER PLATE, Red Anodized Aluminum, 96 Well, PCR Plates, Sculpted to Plate Bottom, use with Heat Blocks, SLAS Footprint
Catalog No. NC9882359 Supplier V&P Scientific Inc Supplier No. VP 741I6A
HEAT TRANSFER PLATE, Red Anodized Aluminum, 96 Well, PCR Plates, Sculpted to Plate Bottom, use with Heat Blocks, SLAS Footprint