Learn More
Braintree Scientific Heat Lamp with base
SDP
Catalog No. 501954937
Encompass
Heat Lamp, HL-1B Arm Length: 2 spring loaded arms each 18 lengths Base: 10" x 10" Weight:17 lbs. Power: 110VAC Bulb: 250 watt infrared heat Lamp Free Standing Infrared heat lamp has an adjustable swing arm providing for easy adjustment of radiant heat to specimen. The lamp is supplied with a 250W(infrared) heat lamp bulb and a built in reflector. A chrome bulb guard is also provided and easily snaps onto the lamp housing after the bulb is installed. Fits standard 110V AC socket. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4937 Supplier Braintree Scientific Supplier No. HL1B120V
Heat Lamp, HL-1B Arm Length: 2 spring loaded arms each 18 lengths Base: 10" x 10" Weight:17 lbs. Power: 110VAC Bulb: 250 watt infrared heat Lamp Free Standing Infrared heat lamp has an adjustable swing arm providing for easy adjustment of radiant heat to specimen. The lamp is supplied with a 250W(infrared) heat lamp bulb and a built in reflector. A chrome bulb guard is also provided and easily snaps onto the lamp housing after the bulb is installed. Fits standard 110V AC socket. **Sold for research purposes only and not for human use**