Learn More
Cole-Parmer HM-200-500-115 Heating Mantle, 500 mL Capacity; 115 VAC
Catalog No. NC0233982 Shop All Cole-Parmer Products
Encompass
Features a durable, chemical resistant polypropylene housing that is "safe to the touch". Flexible heating element ensures minimum risk of flask breakage. These controlled and grounded electromantles feature a built-in power regulator and safety earth (ground) screen. Unique ventilation slots keep the exterior "safe to the touch" even while in use. The stainless steel grounding screen provides added protection against shock in the event of spillage or flask breakage. Mantles include 6 1/2-ft (2-m) power cord; the 115 VAC models include a U.S. plug (230 VAC models do not include plugs).
Catalog No. NC0233982 Supplier Cole-Parmer Supplier No. 3600212
Features a durable, chemical resistant polypropylene housing that is "safe to the touch". Flexible heating element ensures minimum risk of flask breakage. These controlled and grounded electromantles feature a built-in power regulator and safety earth (ground) screen. Unique ventilation slots keep the exterior "safe to the touch" even while in use. The stainless steel grounding screen provides added protection against shock in the event of spillage or flask breakage. Mantles include 6 1/2-ft (2-m) power cord; the 115 VAC models include a U.S. plug (230 VAC models do not include plugs).