Learn More
Heidolph™ Heat-On™ Accessories
GREENER_CHOICESDP
Catalog No. 05404171 Shop All Heidolph North America Products
Change view
Click to view available options
Type:
Adapter Plate
Safety Handle
For Use With (Equipment):
135 mm Hotplate
MR Hei Series Magnetic Stirrer
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
05-404-171 Adapter Plate 135 mm Hotplate
05-404-170 Safety Handle MR Hei Series Magnetic Stirrer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 05-404-171 Supplier Heidolph™ Supplier No. 015882150
Add to Cart
Add to Cart
Accessories for Heat-On™ attachments.
  • Optional safety lifting handles have a simple and secure quick-release mechanism that allows the user to quickly attach the handles and remove the block from the heat source, even whilst it remains hot.
  • Aluminum adapter fits Heat-on™ blocks on 135 mm Hotplates (required for IKA and the Radleys Stirring Hotplate).
  • Specifications
    Description Heat-On Accessory
    For Use With (Equipment) 135 mm Hotplate
    Type Adapter Plate
    WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov