Learn More
New Era Pump Systems Inc Heater control unit & 120V AC 33W, 2 Thermocouplers, 54cm cord, 9cm x 7.5cm syringe heating pad.
Catalog No. NC1042242
Encompass
Heater control unit & 120V AC 33W, 2 Thermocouplers, 54cm cord, 9cm x 7.5cm syringe heating pad.
Catalog No. NC1042242 Supplier New Era Pump Systems Inc Supplier No. HEATERKIT1LG
Heater control unit & 120V AC 33W, 2 Thermocouplers, 54cm cord, 9cm x 7.5cm syringe heating pad.