Learn More
Electron Microscopy Sciences Rinzl 2R High-Heat Plastic Coverslips, 22 x 22 mm
SDP
Catalog No. 501930737
Encompass_Preferred
Plastic coverslips made from clear vinyl plastic. They are unbreakable, and will not scratch or cut fingers. Used in Biological and Medical labs, etc. Thickness of 0.01" or 0.28 mm and refractive index of glass. Economical and half the price of glass. Not autofluorescent.
Size: 22 x 22 mm
Catalog No. 50-193-0737 Supplier Electron Microscopy Sciences Supplier No. 72261232R
Plastic coverslips made from clear vinyl plastic. They are unbreakable, and will not scratch or cut fingers. Used in Biological and Medical labs, etc. Thickness of 0.01" or 0.28 mm and refractive index of glass. Economical and half the price of glass. Not autofluorescent.
Size: 22 x 22 mm