Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Heat and Pour Nutrient Agar
GREENER_CHOICESCI_ED

Catalog No. S01269 Shop All Supertek Scientific Products
Change view
Click to view available options
Quantity:
225 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01269 225 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01269 Supplier Supertek™ Supplier No. CH14034S3
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This 225 mL Heat and Pour Nutrient Agar bottle is very convenient and easy to use.

  • The 225 mL Heat and Pour Nutrient Agar bottle is very convenient and easy to use.
  • Just heat and pour it into your Petri dishes.
  • Allow the agar to set up and you are ready to go.
  • The is enough agar for 10 to 90 mm Petri dishes.
  • Ages 7 plus

Specifications

Form Gel
Manufacturing Origin United States
Product Type Bacterial Growth Medium
Sterility Sterile
Target Organism Class Molds
Test Type Fungal Testing
For Use With (Application) Culture Medium
Quantity 225 mL
Species None
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.