Learn More
MSC Dow Corning 2423472 3 oz Tube Red RTV Silicone Joint Sealant -85 to 500°F Operating Temp, 17 min Tack Free Dry Time, 24 hr Full Cure Time, Series 736
Catalog No. 501113816
Encompass
Dow Corning 2423472 3 oz Tube Red RTV Silicone Joint Sealant -85 to 500°F Operating Temp, 17 min Tack Free Dry Time, 24 hr Full Cure Time, Series 736
Catalog No. 50-111-3816 Supplier MSC Supplier No. 00231860
Dow Corning 2423472 3 oz Tube Red RTV Silicone Joint Sealant -85 to 500°F Operating Temp, 17 min Tack Free Dry Time, 24 hr Full Cure Time, Series 736