Learn More
Alkali Scientific Heat Sealable Blotting Bags, Pre Cut to Mini Gel , Non Protein Binding, 10.2cm x 10.2cm
SDP
Catalog No. 5000662224
Encompass_Preferred
Heat seal the single open end of this non protein binding flexible pouch after inserting the membrane. Manufactured from 100% virgin, high clarity polyethylene film, this pre cut pouch is designed specifically for Western blot membrane handling. The pouch features one open end to easily accept the membrane and three sealed sides that provide a secure enclosure during experimental procedures. It is suitable for use during washing, staining, and de staining steps with either primary or secondary antibodies. The non protein binding surface helps minimize background interaction and supports consistent experimental results. The durable polyethylene film maintains clarity for easy visual inspection and reliable handling in laboratory workflows. Supplied as a convenient pack of 100 pouches for routine laboratory applications and molecular biology workflows.
Catalog No. 50-006-62224 Supplier Alkali Scientific Supplier No. MB9721
Heat seal the single open end of this non protein binding flexible pouch after inserting the membrane. Manufactured from 100% virgin, high clarity polyethylene film, this pre cut pouch is designed specifically for Western blot membrane handling. The pouch features one open end to easily accept the membrane and three sealed sides that provide a secure enclosure during experimental procedures. It is suitable for use during washing, staining, and de staining steps with either primary or secondary antibodies. The non protein binding surface helps minimize background interaction and supports consistent experimental results. The durable polyethylene film maintains clarity for easy visual inspection and reliable handling in laboratory workflows. Supplied as a convenient pack of 100 pouches for routine laboratory applications and molecular biology workflows.