Learn More
Research Products International Corp Heat Sealable Poly Bag, 4 mm Thick, 20 x 25 cm, 100 per Case
SDP
Supplier:  Research Products International Corp 286646C
Encompass
Heat Sealable Polyethylene Bags are transparent and specially designed of use in all hybridization applications. Ideal for quickly sealing solutions with a bag sealer. Bags are 4 mil thick. Poly Bags can be used with an Electronic Bag Sealer also available from RPI.
Catalog No. 50-212-874