Learn More
Cayman Chemical Heat Shock Protein Inhibitor I, Purity: ≥98%, CAS: 218924-25-5, 10 mg
SDP
Catalog No. 502971396
Encompass_Preferred
A benzylidene lactam compound that prevents the synthesis of inducible Hsps Hsp105, Hsp72, and Hsp40; at 100 μM, inhibits the development of thermotolerance in COLO 320 DM cells
Catalog No. 50-297-1396 Supplier Cayman Chemical Supplier No. 1539510 MG
A benzylidene lactam compound that prevents the synthesis of inducible Hsps Hsp105, Hsp72, and Hsp40; at 100 μM, inhibits the development of thermotolerance in COLO 320 DM cells