Learn More
Diagnostic Biosystems HEAT SHOCK PROTEIN 70 HSP 70 W Heat Shock Protein 70 (HSP 70) (W27)
Catalog No. 502407423
Encompass_Preferred
This antibody is specific to constitutive (HSP73) and inducible (HSP72) form of HSP70. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides and other known proteins. HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs and other damaging conditions.
Catalog No. 50-240-7423 Supplier Diagnostic Biosystems Supplier No. PDM597
This antibody is specific to constitutive (HSP73) and inducible (HSP72) form of HSP70. HSP70 is found to be associated with steroid receptors, actin, p53, polyoma T antigen, nucleotides and other known proteins. HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs and other damaging conditions.