Learn More
Cayman Chemical Heat Shock Protein Inhibitor
SDP
Catalog No. 502969036
Encompass_Preferred
A benzylidene lactam compound that prevents the synthesis of inducible Hsps Hsp105, Hsp72, and Hsp40; at 100 μM, inhibits the development of thermotolerance in COLO 320 DM cells
Catalog No. 50-296-9036 Supplier Cayman Chemical Supplier No. 153951 MG
Add to Cart
Add to Cart
A benzylidene lactam compound that prevents the synthesis of inducible Hsps Hsp105, Hsp72, and Hsp40; at 100 μM, inhibits the development of thermotolerance in COLO 320 DM cells