Learn More
Cayman Chemical Heat Shock Protein Inhibitor
SDP
Supplier:  Cayman Chemical 153955 MG
Encompass_Preferred
A benzylidene lactam compound that prevents the synthesis of inducible Hsps Hsp105, Hsp72, and Hsp40; at 100 μM, inhibits the development of thermotolerance in COLO 320 DM cells
Catalog No. 50-296-9071