Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Heat-Shrink Bands for Nalgene™ PETG Media Bottles
GREENER_CHOICE

Catalog No. 0181450 Shop All Thermo Scientific Products
Change view
Click to view available options
Neck Finish:
20-415
24-415
38-430
53B
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish
0181450 20-415
0181451 24-415
0181452 38-430
0181453 53B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0181450 Supplier Thermo Scientific™ Supplier No. 3121600200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ensure the integrity of bottle contents with these tamper-resistant seals. Heat-shrink bands are gamma stable and include a perforated tear strip feature for easy removal.

Ensure the integrity of bottle contents with Thermo Scientific™ Heat-Shrink Bands for Thermo Scientific™ Nalgene™ PETG Media Bottles. Simply apply the recommended application torque for Thermo Scientific Nalgene closures and slide the heat-shrink band over the closure and bottle neck. Heat the band to shrink and secure contents with a tamper-resistant seal.

  • Gamma stable bands include a perforated 'tear strip' feature for easy removal.
  • Bands available are in four sizes to mate with Nalgene PETG Square Media Bottles.
  • Bands are packed 1,000 per case (two zipper bags of 500 each).

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Warranty: 90 days

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Material PETG
No. per Case 1000
No. per Pack 1000
Sterility Non-sterile
Torque 10-14 in.-lb
Torque Wrench Fittings 2195-1020
Neck Finish 20-415
For Use With (Equipment) 30 mL Square Media Bottle
Autoclavable Not Autoclavable
Disposable Single-use
Product Type Heat-shrink Band
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.