Learn More
Cole-Parmer SHM-200-500-115 Stirring/Heating Mantle, 500 mL Capacity; 115 VAC
Catalog No. NC1129577 Shop All Cole-Parmer Products
Encompass
These grounded stirring/heating mantles share all the features of a standard heating mantle plus the added functionality of a magnetic stirrer for a wide range of applications. The stirring circuitry, built into the base of the mantle, provides bidirectional stirring speeds up to 520 rpm. An "auto-recapture" function reactivates stirring if it is interrupted or lost for any reason. Coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support. Built-in power controller has dial markings from 1 to 10 for repeatability.Chemical-resistant polypropylene exterior remains cool to the touch even when the element temperatures reach 842F (450C). Grounded earth screen and double fuses provide addition safety. Two colored lights near the control dial indicate "power on" and "heat on."
Catalog No. NC1129577 Supplier Cole-Parmer Supplier No. 0464130
These grounded stirring/heating mantles share all the features of a standard heating mantle plus the added functionality of a magnetic stirrer for a wide range of applications. The stirring circuitry, built into the base of the mantle, provides bidirectional stirring speeds up to 520 rpm. An "auto-recapture" function reactivates stirring if it is interrupted or lost for any reason. Coiled heating element is suspended within a thermal insulating cartridge to provide maximum heat transfer and support. Built-in power controller has dial markings from 1 to 10 for repeatability.Chemical-resistant polypropylene exterior remains cool to the touch even when the element temperatures reach 842F (450C). Grounded earth screen and double fuses provide addition safety. Two colored lights near the control dial indicate "power on" and "heat on."