Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Heat Transfer Activity Kit
GREENER_CHOICESCI_ED

Catalog No. S139435 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139435 6+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139435 Supplier Supertek™ Supplier No. PH20187
Add to Cart
Add to Cart

The Heat Transder Activity Kit observes heat transfer under different conditions by measuring the temperarture of the insulated containers before and after connecting them with the metal bar.

  • Perfect activity to observe heat transfer under different conditions.
  • Includes...
  • 1 - Plastic cup
  • 1 - Thermocoal cup
  • 2 - Foam Discs
  • 1 - Aluminum Bar
  • 2 - Alcohol thermometers, 10 to 110°C
  • Includes instruction manual

Specifications

Grade 6+
Format Kit
Languages English
Subject Physical Science - Heat
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.