Learn More
Huber USA HEAT TRANSFER FLUID P20.225
Catalog No. NC3962613
Encompass_Preferred
Heat transfer fluid P20.225/275.50emperature range for:- open/closed systems: 20...225°C- Unistats (only closed): 20...275°CMaterial: silicone oilColour: colourless, clearViscosity at 25°C: 50 mm²/sCan: 10 liters
Catalog No. NC3962613 Supplier Huber USA Supplier No. 6158
Heat transfer fluid P20.225/275.50emperature range for:- open/closed systems: 20...225°C- Unistats (only closed): 20...275°CMaterial: silicone oilColour: colourless, clearViscosity at 25°C: 50 mm²/sCan: 10 liters