Learn More
Grainger Wisco Heated Display Case, Two-Door, Stainless Steel, Black, 640 W
Catalog No. 502749461
Encompass_Preferred
This heated display case utilizes circulating, heated air to maintain food items at temperatures above 150°F. Featuring a durable stainless steel base and acrylic panels for optimal visibility, this unit is designed to hold a variety of products in both self-serve and operator-assisted environments.
  • Maintains temperatures ranging from 80°F to over 150°F
  • Adjustable thermostat provides precise temperature control
  • Flexible, adjustable shelving system accommodates diverse food sizes
  • Two-door pass-through configuration for efficient access
  • Constructed with stainless steel corners and base for durability
  • Includes 2 shelves with 14 in. of total vertical shelf space
  • Shatter-resistant light bulb compliant with NSF standards
  • Designed for countertop use
Catalog No. 50-274-9461 Supplier Grainger Supplier No. 14L722
This heated display case utilizes circulating, heated air to maintain food items at temperatures above 150°F. Featuring a durable stainless steel base and acrylic panels for optimal visibility, this unit is designed to hold a variety of products in both self-serve and operator-assisted environments.
  • Maintains temperatures ranging from 80°F to over 150°F
  • Adjustable thermostat provides precise temperature control
  • Flexible, adjustable shelving system accommodates diverse food sizes
  • Two-door pass-through configuration for efficient access
  • Constructed with stainless steel corners and base for durability
  • Includes 2 shelves with 14 in. of total vertical shelf space
  • Shatter-resistant light bulb compliant with NSF standards
  • Designed for countertop use