Learn More
Braintree Scientific Heated Hard Pad Connectors for Androit Pump
SDP
Catalog No. 501954935
Encompass
Water Heated Hard Pads - Connects to Gaymar, Androit and other pumps. Durable polycarbonate construction. Scalpel, needle, and towel clamp proof. Smooth surfaces are easy to clean helping to prevent nosocomial infection. Highly puncture resistant. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4935 Supplier Braintree Scientific Supplier No. HHPANDROIT
Water Heated Hard Pads - Connects to Gaymar, Androit and other pumps. Durable polycarbonate construction. Scalpel, needle, and towel clamp proof. Smooth surfaces are easy to clean helping to prevent nosocomial infection. Highly puncture resistant. **Sold for research purposes only and not for human use**