Learn More
Glas Col LLC Heating Mantle
SDP
Catalog No. NC1096020
Encompass_Preferred
Fabric heating mantle for 60 degree funnel with 6" top diameter. This mantle reduces viscosity and keeps solutions warm during transfer.Heats to 400 degrees C maximum internal operating temperature. 120W, 115V, CSA.
Catalog No. NC1096020 Supplier Glas Col LLC Supplier No. 100A O718
Fabric heating mantle for 60 degree funnel with 6" top diameter. This mantle reduces viscosity and keeps solutions warm during transfer.Heats to 400 degrees C maximum internal operating temperature. 120W, 115V, CSA.