Learn More
Leica Biosystems HistoCore Arcadia H Heated Paraffin Embedding Station - Leica Biosystems REF 14039357258
Catalog No. NC3566301
Encompass
The HistoCore Arcadia H heated paraffin embedding station provides precise temperature control, ergonomic specimen handling, and an intuitive touchscreen interface to streamline routine and high-throughput embedding workflows in histology laboratories.
  • Ergonomic design with wristrests and a magnifier for precise specimen orientation.
  • Intuitive LCD touchscreen for easy control and monitoring.
  • Adjustable temperature control (50°C-75°C) for reproducible embedding.
  • Large removable cassette tray holds approximately 150 cassettes for high throughput.
  • 4 L paraffin tank capacity reduces refill frequency.
  • Compact footprint (636 x 560 x 384 mm) and 27 kg weight for benchtop use.
Catalog No. NC3566301 Supplier Leica Biosystems Supplier No. 14039357258
Add to Cart
Add to Cart
Request a Quote
The HistoCore Arcadia H heated paraffin embedding station provides precise temperature control, ergonomic specimen handling, and an intuitive touchscreen interface to streamline routine and high-throughput embedding workflows in histology laboratories.
  • Ergonomic design with wristrests and a magnifier for precise specimen orientation.
  • Intuitive LCD touchscreen for easy control and monitoring.
  • Adjustable temperature control (50°C-75°C) for reproducible embedding.
  • Large removable cassette tray holds approximately 150 cassettes for high throughput.
  • 4 L paraffin tank capacity reduces refill frequency.
  • Compact footprint (636 x 560 x 384 mm) and 27 kg weight for benchtop use.