Learn More
Euthanex Heated Regulator (0-100 PSI Output), connects directly to the CO2 cylinder with CGA-320 fitting.
Catalog No. NC1880331
Encompass
Heated Regulator (0-100 PSI Output), connects directly to the CO2 cylinder with CGA-320 fitting. Capable of handling high flow rates. Incorporated electric heater keeps unit from freezing during operation. Standard 120 VAC. Includes 9' connecting hose with quick-disconnect fitting.
Catalog No. NC1880331 Supplier Euthanex Supplier No. EA550
Heated Regulator (0-100 PSI Output), connects directly to the CO2 cylinder with CGA-320 fitting. Capable of handling high flow rates. Incorporated electric heater keeps unit from freezing during operation. Standard 120 VAC. Includes 9' connecting hose with quick-disconnect fitting.