Learn More
Electron Microscopy Sciences Tripod Polisher Heater Block
SDP
Catalog No. 50980874
Encompass
Designed to hold any of the Lbrackets while being heated on a hot plate prior to mounting sample melted wax. Provides a slow, uniform temperature gradient, which minimizes thermal shock. - For use with Tripod Polisher.
Catalog No. 50-980-874 Supplier Electron Microscopy Sciences Supplier No. 5011650
Designed to hold any of the Lbrackets while being heated on a hot plate prior to mounting sample melted wax. Provides a slow, uniform temperature gradient, which minimizes thermal shock. - For use with Tripod Polisher.