Learn More
Warner Instruments™ Heater Elements for Warner Chambers and Platforms, 2 Pk.
Shop All BTX Harvard Products
Description
- RH-2
- Replacement heater blocks for Series PH and Series PM platforms Legacy non-heater platforms can be converted to heater platforms with the RH-2 upgrade kit consisting of a pair of resistive heater blocks, heat transfer pads, and mounting screws. Tapped holes for the heater blocks are included in all plain platforms. The only tool required for installation is a small Philips screwdriver. Note: Very old platform models may not be upgradable. Consult Warner Technical Support if your platform does not have slots under the wings.
Specifications
Specifications
| For Use With (Equipment) | Warner Series PH and Series PM Platforms |
| Type | Replacement Heating Element |
| Quantity | 2/Pk. |