Learn More
Waters Corp Heater-Shaker, Deepwell Plate Adaptor, High-speed Mixing, Temperature Control
Catalog No. 502391639 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Provides high-speed mixing action & temperature control for the most demanding applications. This uniquely designed compact device, which is suitable for a wide range of labware, combines both heating, from ambient to 99°C, and vibration-free shaking action from 200 to 2,500 rpm.
  • High-speed mixing action
  • Precise temperature control from ambient to 99°C
  • Vibration-free shaking action from 200 to 2,500 rpm
  • Compact device
  • Compatible with a wide range of labware
  • System compatibility with Andrew+, Pipette+, and Manual systems
  • Software compatibility with OneLab
Catalog No. 50-239-1639 Supplier Waters Corp Supplier No. 186010426
Add to Cart
Add to Cart
Request a Quote
Provides high-speed mixing action & temperature control for the most demanding applications. This uniquely designed compact device, which is suitable for a wide range of labware, combines both heating, from ambient to 99°C, and vibration-free shaking action from 200 to 2,500 rpm.
  • High-speed mixing action
  • Precise temperature control from ambient to 99°C
  • Vibration-free shaking action from 200 to 2,500 rpm
  • Compact device
  • Compatible with a wide range of labware
  • System compatibility with Andrew+, Pipette+, and Manual systems
  • Software compatibility with OneLab