Learn More
Waters Corp Heater-Shaker, HP Plate Adaptor, 200 to 2,500 rpm, Ambient to 99°C
Catalog No. 502391944 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Heater-Shaker offers high-speed mixing and temperature control for demanding applications. This compact device combines heating, from ambient to 99°C, with vibration-free shaking action ranging from 200 to 2,500 rpm.
  • High-speed mixing action
  • Temperature control from ambient to 99°C
  • Vibration-free shaking from 200 to 2,500 rpm
  • Compact design
  • System compatibility with Andrew+, Manual, Pipette+
  • Software compatibility with OneLab
Catalog No. 50-239-1944 Supplier Waters Corp Supplier No. 176005080
Add to Cart
Add to Cart
Request a Quote
The Heater-Shaker offers high-speed mixing and temperature control for demanding applications. This compact device combines heating, from ambient to 99°C, with vibration-free shaking action ranging from 200 to 2,500 rpm.
  • High-speed mixing action
  • Temperature control from ambient to 99°C
  • Vibration-free shaking from 200 to 2,500 rpm
  • Compact design
  • System compatibility with Andrew+, Manual, Pipette+
  • Software compatibility with OneLab